missing translation for 'onlineSavingsMsg'
Learn More

Tridodecylamine 85.0+%, TCI America™

Catalog No. T351925ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T351925ML 25 mL
T3519100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T351925ML Supplier TCI America Supplier No. T351925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-87-4
Molecular Formula C36H75N
Molecular Weight (g/mol) 522.003
MDL Number MFCD00008971
InChI Key SWZDQOUHBYYPJD-UHFFFAOYSA-N
Synonym tridodecylamine, trilaurylamine, tri-n-dodecylamine, alamine 304, tridodecyl amine, adogen 360, 1-dodecanamine, n,n-didodecyl, n,n-didodceyl-1-dodecanamine, n,n-didodecyl-1-dodecanamine, unii-sf36t21u17
PubChem CID 7624
IUPAC Name N,N-didodecyldodecan-1-amine
SMILES CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 296°C
Quantity 25 mL
Formula Weight 522.00
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridodecylamine
EINECSNumber (2)-0176
TSCA Yes