missing translation for 'onlineSavingsMsg'
Learn More

Tridecanal 95.0+%, TCI America™

Catalog No. T04105ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04105ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T04105ML Supplier TCI America Supplier No. T04105ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10486-19-8
Molecular Formula C13H26O
Molecular Weight (g/mol) 198.35
MDL Number MFCD00007018
InChI Key BGEHHAVMRVXCGR-UHFFFAOYSA-N
Synonym tridecyl aldehyde, tridecanaldehyde, 1-tridecanal, n-tridecylaldehyde, n-tridecanal, tridecane aldehyde, tridecanealdehyde, aldehyde c-13, technical, tridecanal
PubChem CID 25311
IUPAC Name tridecanal
SMILES CCCCCCCCCCCCC=O

Specifications

Melting Point 14°C
Color Yellow
Boiling Point 280°C
Quantity 5 mL
Formula Weight 198.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecanal
EINECSNumber (2)-0494
TSCA Yes
Recommended Storage Refrigerator