missing translation for 'onlineSavingsMsg'
Learn More

Tricosane 99.0+%, TCI America™

Catalog No. S02951G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02951G Supplier TCI America Supplier No. S02951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 638-67-5
Molecular Formula C23H48
Molecular Weight (g/mol) 324.637
MDL Number MFCD00009350
InChI Key FIGVVZUWCLSUEI-UHFFFAOYSA-N
Synonym n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
PubChem CID 12534
ChEBI CHEBI:32934
IUPAC Name tricosane
SMILES CCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 47°C
Color White
Boiling Point 234°C
Quantity 1 g
Formula Weight 324.64
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tricosane [Standard Material for GC]
EINECSNumber (2)-0010
TSCA Yes