Learn More
Cytiva Tricorn Columns
Tricorn columns are compatible with high flow rates and are designed for high-performance media such as Sepharose, Superdex, and SOURCE.
$1174.36 - $2620.13
Specifications
| Product Type | Tricorn Columns (Empty) |
|---|---|
| Quantity | 1 Ea. |
| Includes | Glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock and filter holder |
| Material | Borosilicate glass |
| Support Type | Polypropylene filter |
| Catalog Number | Mfr. No. | Diameter (Metric) Inner | Height (Metric) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) Inner | Height (Metric) | Price | Quantity | ||||
|
45-200-35
|
Cytiva
28406408 |
5 mm | 20 mm |
Each for $1,174.36
|
|
||||
|
45-200-34
|
Cytiva
28406411 |
5 mm | 150 mm |
Each for $1,516.92
|
|
||||
|
45-200-36
|
Cytiva
28406413 |
10 mm | 20 mm |
Each for $1,524.40
|
|
||||
|
45-200-37
|
Cytiva
28406419 |
10 mm | 60 mm |
Each for $2,620.13
|
|
||||
|
45-200-38
|
Cytiva
28406412 |
5 mm | 200 mm |
Each for $1,764.55
|
|
||||
|
45-200-42
|
Cytiva
28406417 |
10 mm | 200 mm |
Each for $1,762.05
|
|
||||
|
4520040
|
Cytiva
28406414 |
10 mm | 50 mm |
Each for $1,593.05
|
|
||||
|
45-200-41
|
Cytiva
28406416 |
10 mm | 150 mm |
Each for $1,593.90
|
|
||||
|
45-200-43
|
Cytiva
28406418 |
10 mm | 300 mm |
Each for $1,903.97
|
|
||||
|
45-200-45
|
Cytiva
28406415 |
10 mm | 100 mm |
Each for $1,593.90
|
|
||||
|
45-200-44
|
Cytiva
28406409 |
5 mm | 50 mm |
Each for $1,321.64
|
|
||||
|
45-200-39
|
Cytiva
28406410 |
5 mm | 100 mm |
Each for $1,494.49
|
|
||||
Description
- Tricorn empty columns are designed for high performance separations on small and medium scale
- The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
- Broad range of column dimensions suitable for many different purification needs.
- Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
- Separate packing connector allows simple column packing for a broad range of media.
- Locking ring prevents accidental compression of media bed.
Specifications
| Tricorn Columns (Empty) | |
| Glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock and filter holder | |
| Polypropylene filter |
| 1 Ea. | |
| Borosilicate glass | |
| High resolution liquid chromatography of biomolecules performed at medium to high pressures |