missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, MP Biomedicals

5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer

$192.49 - $1137.40

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN807413
View Documents
MP Biomedicals Inc
04807413
100 g
Each for $192.49
Only null left
 
ICN807416
View Documents
MP Biomedicals Inc
04807416
250 g
Each for $501.32
Only null left
 
ICN807420
View Documents
MP Biomedicals Inc
04807420
1 kg
Each for $1,137.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4-1-5704
179.17
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C6H13NO5
SEQKRHFRPICQDD-UHFFFAOYSA-N
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO

Specifications

4-1-5704
C6H13NO5
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
Tricine
100 g
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
Electrophoresis
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'