missing translation for 'onlineSavingsMsg'
Learn More

Tricine, Electrophoresis Grade, MP Biomedicals

5704-04-1 Purity: >>>99% Heavy metals (as Pb): <<10 ppm pKa at 25°C = 8.1 Biological buffer

Supplier:  MP Biomedicals Inc 04807416

 View more versions of this product

Catalog No. ICN807416


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tricine
250 g
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
Electrophoresis
4-1-5704
C6H13NO5
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063

Chemical Identifiers

4-1-5704
179.17
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C6H13NO5
SEQKRHFRPICQDD-UHFFFAOYSA-N
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions