missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetyl chloride, 99%

Catalog No. AAB2167530
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2167530 250 g
AAB2167518 50 g
AAB216750B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2167530 Supplier Thermo Scientific Chemicals Supplier No. B2167530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76-02-8 | C2Cl4O | 181.821 g/mol

Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.

Solubility
Reacts violently with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, alcohols, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 76-02-8
Molecular Formula C2Cl4O
Molecular Weight (g/mol) 181.821
MDL Number MFCD00000792
InChI Key PVFOMCVHYWHZJE-UHFFFAOYSA-N
Synonym trichloroacetyl chloride, superpalite, acetyl chloride, trichloro, trichloroacetochloride, trichloroacetic acid chloride, ccl3cocl, trichloroacetylchloride, unii-9sn86t76y6, ccris 6764, acetyl chloride, 2,2,2-trichloro
PubChem CID 6420
IUPAC Name 2,2,2-trichloroacetyl chloride
SMILES C(=O)(C(Cl)(Cl)Cl)Cl

Specifications

Melting Point -146°C
Density 1.629
Boiling Point 114°C to 116°C
Refractive Index 1.47
Quantity 250 g
UN Number UN2442
Beilstein 774120
Sensitivity Moisture sensitive
Solubility Information Reacts violently with water.
Formula Weight 181.83
Percent Purity 99%
Chemical Name or Material Trichloroacetyl chloride
Show More Show Less
Hazard Category H302-H314-H330-H335
Hazard Statement GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TRICHLOROACETYL CHLORIDE
EINECSNumber 200-926-7
RTECSNumber AO7140000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only