missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid Solution, MP Biomedicals™

Catalog No. ICN19605780
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19605780 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19605780 Supplier MP Biomedicals Inc Supplier No. 0219605780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trichloroacetic acid Solution

  • It is used in protein precipitation; has been used to determine protein concentration by quantitative precipitation.
  • It is also Used as a decalcifier and fixative in microscopy.
  • A protocol for the precipitation of nucleic acids can be found in Molecular Cloning. Rats exposed to chronic sublethal amounts of trichloroacetic acid displayed an increase in serum bilirubin with a decrease in hematological proteins and cholesterols along with significant decreases in red blood corpuscles, mean cell volume, mean corpuscular hemoglobin, hemoglobin and hematocrit.
  • In mice the production of liver tumors was associated to Trichloroacetic acid, which is also a known mouse hepatocarinogen.
  • On the guinea pig trichloroacetic acid was observed to be a mild alergen on the skin.
  • Trichloroacetic acid is an acetic acid analogue commonly used to precipitate proteins, DNA and RNA. In the presence of SDS Trichloroacetic acid will precipitate proteins which can then be quantified by the Lowry method.
  • The compound has also found uses as a decalcifier and fixative in microscopy, in protein sequencing, detecting albumin and organic synthesis.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.4
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

pH 1.2 (0.1 M aqueous solution)(Literature)
Melting Point ∼56°C (Literature)
Concentration 100% w/v
Density 1.62 sp. gr.
Quantity 100 mL
Physical Form Solution
UN Number UN2564
Beilstein 970119
Formula Weight 163.38
Chemical Name or Material Trichloroacetic Acid Solution, 6.1N
RTECSNumber AJ7875000
storageNote1 +4°C