missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic Acid, 15% w/v (AOAC), Fisher Chemical

Catalog No. SA94151
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA94151 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA94151 Supplier Ricca Chemical Company Supplier No. SA94151

Chemical Identifiers

CAS 7732-18-5 , 76-03-9 , 7440-44-0
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) Mixture
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

CAS Min % 12.77, 85.0, 0.17
CAS Max % 13.29, 89.0, 0.18
Percent Purity 15% w/v
Grade Laboratory
Quantity 1 L
Packaging Amber Glass Bottle
Chemical Name or Material Trichloroacetic Acid, 15% (w/v)