missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Trichloro(1H,1H,2H,2H-perfluorooctyl)silane

Catalog No. 44893110G
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44893110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44893110G Supplier Sigma Aldrich Supplier No. 44893110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichloro(1H,1H,2H,2H-perfluorooctyl)silane (PFOCTS) is a chlorosilane that forms a self-assembled monolayer (SAM) on a variety of substrates. It is majorly used to modify the surface with superhydrophobic properties. It is a biocompatible polymer with low surface energy. PFOCTS exhibits anti-fouling properties with high wettability which facilitates the release of different composites from the mold.PFOCTS is a coupling agent that is used to silanize the substrate for the fabrication of microfluidic devices using soft lithography. It can also be used on silicon base that forms a chemically patterned surface for quantitative analysis of insoluble surfactants in the polymeric matrix. Alumina membranes can be treated with PFOCTS for the formation of surperhydrophobic polyimide (PI) based nanotube arrays.

Specifications

CAS 78560-45-9
Density 1.3 g/mL (at 25°C)
Boiling Point 192°C
Molecular Formula C8H4Cl3F13Si
Refractive Index n20/D 1.352 (literature)
Linear Formula CF3(CF2)5 CH2CH2SiCl3
Quantity 10 g
MDL Number MFCD00042363
Synonym Trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane; 1 H,1 H,2 H,2 H-Perfluorooctyl-trichlorosilane
Molecular Weight (g/mol) 481.54
Percent Purity 97%
Show More Show Less