missing translation for 'onlineSavingsMsg'
Learn More

Tricarbonyl(cyclooctatetraene)iron 96.0+%, TCI America™

Catalog No. T17751G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17751G Supplier TCI America Supplier No. T17751G

Chemical Identifiers

CAS 12093-05-9
Molecular Formula C11H8FeO3
Molecular Weight (g/mol) 244.027
MDL Number MFCD00001755
InChI Key XAOCYVWRKJTXOL-JGZYGLCTSA-N
Synonym cyclooctatetraene iron tricarbonyl, tricarbonyl cyclooctatetraene iron, cyclooctatetraenetricarbonyliron, tricarbonyl cyclooctatetraene iron ii, 1,3,5,7-cyclooctatetraeneiron tricarbonyl, tris carbon monoxide cyclooctatetraene iron
PubChem CID 11436476
IUPAC Name carbon monoxide;cyclooctatetraene;iron
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].C1=CC=CC=CC=C1.[Fe]

Specifications

Color Purple-Red
Quantity 1 g
UN Number 3466
Formula Weight 244.03
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tricarbonyl(cyclooctatetraene)iron