missing translation for 'onlineSavingsMsg'
Learn More

Tributylsilane 98.0+%, TCI America™

Numéro de catalogue. p-7137077
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T20435G 5 g
T204325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T20435G Fournisseur TCI America Code fournisseur T20435G
Il en reste null

Identifiants chimiques

CAS 998-41-4
Molecular Formula C12H27Si
Molecular Weight (g/mol) 199.43
MDL Number MFCD00009415
InChI Key ISEIIPDWJVGTQS-UHFFFAOYSA-N
PubChem CID 6327711
IUPAC Name tributylsilyl
SMILES CCCC[Si](CCCC)CCCC

Spécifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
Formula Weight 200.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributylsilane