missing translation for 'onlineSavingsMsg'
Learn More

Tributyl Phosphite 93.0+%, TCI America™

Catalog No. T0362500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0362500ML 500 mL
T036225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0362500ML Supplier TCI America Supplier No. T0362500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-85-2
Molecular Formula C12H27O3P
Molecular Weight (g/mol) 250.319
MDL Number MFCD00009437
InChI Key XTTGYFREQJCEML-UHFFFAOYSA-N
Synonym tri-n-butyl phosphite, phosphorous acid, tributyl ester, tributoxyphosphine, tributylfosfit, butyl phosphite c4h9o 3p, tributylfosfit czech, tributylphosphite, phosphorus tributoxide p obu 3, tri-n-butylphosphite, 1-butanol, 1,1',1-phosphinidynetri
PubChem CID 7623
IUPAC Name tributyl phosphite
SMILES CCCCOP(OCCCC)OCCCC

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 125°C
Quantity 500 mL
Formula Weight 250.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributyl Phosphite
EINECSNumber (2)-2906
RTECSNumber TH0875000
TSCA Yes