missing translation for 'onlineSavingsMsg'
Learn More

Tributyl 2-acetylcitrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015527
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015527 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015527 Supplier Merck Emd Millipore Supplier No. PHR11561G
Tributyl 2-acetylcitrate is a hydrophobic biodegradable plasticizer, that is usually synthesized by esterification of citric acid. Pharmaceutically, it is used as an excipient, which is generally applied as a thin film over the tablets or capsules.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 77-90-7
Vapor Pressure 0.26 psi (20°C)
Density 1.05 g/mL (at 25°C)
Flash Point 217°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CH3 CO2 C[CO2(CH2)3 CH3 ][CH2 CO2(CH2)3 CH3]2
MDL Number MFCD00043554
Synonym Acetyl tributyl citrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 402.48
Grade Certified Reference Material
Show More Show Less