missing translation for 'onlineSavingsMsg'
Learn More

Triamylamine 98.0+%, TCI America™

Catalog No. T117725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T117725ML 25 mL
T1177500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T117725ML Supplier TCI America Supplier No. T117725ML

Chemical Identifiers

CAS 621-77-2
Molecular Formula C15H33N
Molecular Weight (g/mol) 227.436
MDL Number MFCD00015270
InChI Key OOHAUGDGCWURIT-UHFFFAOYSA-N
Synonym triamylamine, tripentylamine, tri-n-pentylamine, 1-pentanamine, n,n-dipentyl, tri-n-amylamine, ccris 4877, tripentyl amine, 1-pentanamine,n-dipentyl, acmc-209n0e, 1-pentanamine,n,n-dipentyl
PubChem CID 12133
IUPAC Name N,N-dipentylpentan-1-amine
SMILES CCCCCN(CCCCC)CCCCC

Specifications

Color Yellow
Boiling Point 240°C
Quantity 25 mL
UN Number 2735
Formula Weight 227.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triamylamine
EINECSNumber (2)-3526
TSCA Yes