missing translation for 'onlineSavingsMsg'
Learn More

Triamyl Phosphate 98.0+%, TCI America™

Catalog No. P02655ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02655ML 5 mL
P026525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02655ML Supplier TCI America Supplier No. P02655ML
Only null left

Chemical Identifiers

CAS 2528-38-3
Molecular Formula C15H33O4P
Molecular Weight (g/mol) 308.399
MDL Number MFCD00015348
InChI Key QJAVUVZBMMXBRO-UHFFFAOYSA-N
Synonym Phosphoric Acid Triamyl Ester, Tripentyl Phosphate
PubChem CID 75665
IUPAC Name tripentyl phosphate
SMILES CCCCCOP(=O)(OCCCCC)OCCCCC

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 mL
Formula Weight 308.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triamyl Phosphate
EINECSNumber (2)-2000
RTECSNumber TC8312500
TSCA No