missing translation for 'onlineSavingsMsg'
Learn More

Triammonium 4-Sulfophthalate (contains 3-Sulfophthalate) 65.0+%, TCI America™

Catalog No. S031525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S031525G Supplier TCI America Supplier No. S031525G
Only null left

Chemical Identifiers

CAS 22411-24-1
Molecular Formula C8H15N3O7S
Molecular Weight (g/mol) 297.28
MDL Number MFCD00134235
InChI Key RMPQIFXEQFYSEM-UHFFFAOYSA-N
Synonym 4-Sulfophthalic Acid Triammonium Salt
PubChem CID 122359803
IUPAC Name 4-sulfobenzene-1,2-dicarboxylic acid triamine
SMILES N.N.N.OC(=O)C1=C(C=C(C=C1)S(O)(=O)=O)C(O)=O

Specifications

Quantity 25 g
Formula Weight 297.28
Percent Purity ≥65.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Triammonium 4-Sulfophthalate (contains 3-Sulfophthalate)
EINECSNumber (3)-2131
TSCA No