missing translation for 'onlineSavingsMsg'
Learn More

Triallyl Isocyanurate (stabilized with BHT) 96.0+%, TCI America™

Catalog No. I027925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I027925G 25 g
I0279100G 100 g
I0279500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I027925G Supplier TCI America Supplier No. I027925G

Chemical Identifiers

CAS 1025-15-6
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00006554
InChI Key KOMNUTZXSVSERR-UHFFFAOYSA-N
Synonym triallyl isocyanurate, isocyanuric acid triallyl ester, 1,3,5-triallyl-1,3,5-triazinane-2,4,6-trione, triallylisocyanurate, taic, diak 7, 1,3,5-triallylisocyanurate, 1,3,5-triallyl isocyanurate, 1,3,5-triallylisocyanuric acid, 1,3,5-tri-2-propenyl-1,3,5-triazine-2,4,6 1h,3h,5h-trione
PubChem CID 13931
IUPAC Name 1,3,5-tris(prop-2-enyl)-1,3,5-triazinane-2,4,6-trione
SMILES C=CCN1C(=O)N(C(=O)N(C1=O)CC=C)CC=C

Specifications

Color White-Yellow
Boiling Point 140°C
Quantity 25 g
Formula Weight 249.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Triallyl Isocyanurate (stabilized with BHT)
EINECSNumber (5)-1047
RTECSNumber XZ1915000
TSCA Yes