missing translation for 'onlineSavingsMsg'
Learn More

Triacetylresveratrol 98.0+%, TCI America™

Catalog No. T32325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32325G 5 g
T32321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32325G Supplier TCI America Supplier No. T32325G
Only null left

Chemical Identifiers

CAS 42206-94-0
Molecular Formula C20H18O6
Molecular Weight (g/mol) 354.358
MDL Number MFCD01546481
InChI Key PDAYUJSOJIMKIS-SNAWJCMRSA-N
PubChem CID 5962587
IUPAC Name [4-[(E)-2-(3,5-diacetyloxyphenyl)ethenyl]phenyl] acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=CC2=CC(=CC(=C2)OC(=O)C)OC(=O)C

Specifications

Melting Point 119°C
Color White
Quantity 5 g
Synonym 3,4′,5-Triacetoxy-trans-stilbene
Formula Weight 354.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triacetylresveratrol