missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-O-benzyl-D-glucal 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 57°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 416.52 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tri-O-benzyl-D-glucal |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |