missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin methoxide, 97%

Catalog No. AAA1335530
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1335530 250 g
AAA1335522 100 g
AAA133550B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1335530 Supplier Thermo Scientific Chemicals Supplier No. A1335530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 1067-52-3 | C13H30OSn | 321.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1067-52-3
Molecular Formula C13H30OSn
Molecular Weight (g/mol) 321.092
MDL Number MFCD00009419
InChI Key KJGLZJQPMKQFIK-UHFFFAOYSA-N
Synonym tributyltin methoxide, stannane, tributylmethoxy, unii-9cff2afn2v, tri-n-butyltin methanolate, 9cff2afn2v, methoxytributyltin, tin, tributylmethoxy, tributyltin methanolate, stannane,tributylmethoxy, stannane, methoxytributyl
PubChem CID 16683411
IUPAC Name tributyl(methoxy)stannane
SMILES CCCC[Sn](CCCC)(CCCC)OC

Specifications

Density 1.12
Boiling Point 124°C to 126°C (4 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.474
Quantity 250 g
UN Number UN2788
Beilstein 3600960
Sensitivity Moisture sensitive
Formula Weight 321.09
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin methoxide
Show More Show Less
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 213-933-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only