missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl borate, 98%

Catalog No. AAA19322AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA19322AP 500 mL
AAA19322AE 100 mL
AAA193220F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA19322AP Supplier Thermo Scientific Chemicals Supplier No. A19322AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 688-74-4 | C12H27BO3 | 230.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 688-74-4
Molecular Formula C12H27BO3
Molecular Weight (g/mol) 230.155
MDL Number MFCD00009434
InChI Key LGQXXHMEBUOXRP-UHFFFAOYSA-N
Synonym n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
PubChem CID 12712
IUPAC Name tributyl borate
SMILES B(OCCCC)(OCCCC)OCCCC

Specifications

Melting Point <-70°C
Density 0.853
Boiling Point 230°C to 235°C
Flash Point 93°C (199°F)
Refractive Index 1.4095
Quantity 500 mL
Beilstein 1703865
Sensitivity Moisture sensitive
Formula Weight 230.16
Percent Purity 98%
Chemical Name or Material Tri-n-butyl borate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-706-5
RTECSNumber ED4900000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only