missing translation for 'onlineSavingsMsg'
Learn More

Tri(2-furyl)phosphine, 97%

Catalog No. AAL13329MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13329MD 250 mg
AAL1332903 1 g
AAL1332906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13329MD Supplier Thermo Scientific Chemicals Supplier No. L13329MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5518-52-5 | C12H9O3P | 232.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5518-52-5
Molecular Formula C12H9O3P
Molecular Weight (g/mol) 232.18
MDL Number MFCD00151857
InChI Key DLQYXUGCCKQSRJ-UHFFFAOYSA-N
Synonym tri 2-furyl phosphine, tri furan-2-yl phosphine, tri-2-furyl phosphine, tris furan-2-yl phosphane, tri-2-furylphosphine, tris 2-furyl phosphine, tri furan-2-yl phosphane, trifurylphosphine, tri fur-2-yl phosphane, tris 2-furanyl phosphine
PubChem CID 521585
IUPAC Name tris(furan-2-yl)phosphane
SMILES O1C=CC=C1P(C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 59°C to 64°C
Boiling Point 136°C (4 mmHg)
Quantity 250 mg
Beilstein 1577564
Formula Weight 232.18
Percent Purity 97%
Physical Form Crystals or Powder or Crystalline Powder or Fused Solid
Chemical Name or Material Tri(2-furyl)phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only