missing translation for 'onlineSavingsMsg'
Learn More

Trehalose Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014891
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014891 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014891 Supplier Merck Emd Millipore Supplier No. PHR1344500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trehalose is a naturally occurring disaccharide which is used as a nontoxic cryoprotectant of enzymes, membranes and vaccines. It serves as a carbohydrate reserve in microorganisms and protects them from adverse conditions.

Specifications

CAS 6138-23-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C12H22O11 · 2H2 O
MDL Number MFCD00071594
Synonym D-(+)-Trehalose dihydrate; alpha,alpha-Trehalose; alpha-D-Glucopyranosyl-alpha-D-glucopyranoside
Type Certified Reference Material
Quantity 500 mg
Formula Weight 378.33
Grade Certified Reference Material