missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Trehalose, Dihydrate

Non-reducing disaccharide which yields 2 moles of D-glucose upon acid hydrolysis. Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro.

$141.00 - $1,810.00

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
Synonym 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
62562550GM
View Documents Promotion Details
MilliporeSigma™
62562550GM
50 g
Each for $141.00
Only null left
 
6256251KG
View Documents Promotion Details
MilliporeSigma™
6256251KG
1 kg
Each for $1,810.00
Only null left
 
Description

Description

  • CAS Number: 6138-23-4
  • Form: Solid
  • Synonyms: 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
  • Formula Weight: 378.3g/mol
Specifications

Chemical Identifiers

6138-23-4
378.33
DPVHGFAJLZWDOC-DJCWUSJTNA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
C12H26O13
MFCD00071594
1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

6138-23-4
MFCD00071594
DPVHGFAJLZWDOC-DJCWUSJTNA-N
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
378.3g/mol
C12H26O13
1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
378.33
Videos
SDS
Documents

Documents