missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Trehalose, Dihydrate

Catalog No. 62562550GM
Change view
Click to view available options
Quantity:
50 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62562550GM 50 g
6256251KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 62562550GM Supplier MilliporeSigma™ Supplier No. 62562550GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Non-reducing disaccharide which yields 2 moles of D-glucose upon acid hydrolysis. Inhibits polyglutamine-mediated protein aggregation both in vivo and in vitro.

  • CAS Number: 6138-23-4
  • Form: Solid
  • Synonyms: 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
  • Formula Weight: 378.3g/mol

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H26O13
Molecular Weight (g/mol) 378.33
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-DJCWUSJTNA-N
Synonym 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
SMILES O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 50 g
Formula Weight 378.3g/mol
Percent Purity ≥97.5% (Assay)
Physical Form Solid