Learn More
METTLER TOLEDO™ TranspBox Accessory for Automatic Dosing
Description
- One-step, safe and accurate dispensing of very small samples
- Compatible with XPR analytical balances
- Minimizes sample loss
- Eliminates transfer errors
Spécifications
Spécifications
| For Use With (Equipment) | Excellence XPR Automatic Balance |
| Size (Metric) | 16 mL |
| Quantity | 1 Ea. |
| Product Line | TranspBox with Vial |
| Type | TranspBox with Vial |