missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Transparent Back Panel For Captair 391 Fume Hood
GREENER_CHOICE

Catalog No. 08444013
Change view
Click to view available options
Material:
Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
08444013 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08444013 Supplier Erlab™ Supplier No. 391EDU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Clear back panel made of synthetic glass offering 360° visibility for 391 fume hood models.

  • For use with Captair 391 Ductless Filtering Fume Hood
  • Includes warranty
  • Useful for teaching and student observation

Specifications

For Use With (Equipment) Captair Ductless Filtering Fume Hoods or CaptairFlow Laminar Flow Hoods
Height (English) 3 in.
Weight (English) 18 lb.
Weight (Metric) 8.2 Kg
Height (Metric) 76.2 mm
Includes Educational Back Panel and Thumb Screws
Product Type Back Panel
Voltage 0/1 V
Material Acrylic
For Use With (Application) Teaching Observation or Locating in Front of Window
Color Clear
Cut Style Rounded Edge
Warranty One Year
Show More Show Less