Learn More
LGC Genomics Llc (Lucigen) TransforMax EPI300-T1R Electrocompetent E. coli, 20 RxnsClone large inserts in standard plasmids, avoid T1 and T5 phage contamination, and control copy number of CopyControl vectors.
Numéro de catalogue. NC2162043
Encompass
Standard cloning for large inserts and T1 phage resistanceAvoid phage contamination with tonA mutation for resistance to bacteriophages T1 and T5.Clone large inserts and transform large plasmids - up to at least 145 kb plasmid DNA.Generate clones with inducible copy number using Copy Control vectors.Achieve high transformation efficienciesTransforMax EPI300-T1R Electrocompetent E. coli are supplied as 10 x 100 L aliquots, with CopyControl™ Induction Solution and pUC19 Control DNA (100 pg/L).
Numéro de catalogue. NC2162043 Fournisseur LGC Genomics Llc Code fournisseur EC02T110
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Standard cloning for large inserts and T1 phage resistanceAvoid phage contamination with tonA mutation for resistance to bacteriophages T1 and T5.Clone large inserts and transform large plasmids - up to at least 145 kb plasmid DNA.Generate clones with inducible copy number using Copy Control vectors.Achieve high transformation efficienciesTransforMax EPI300-T1R Electrocompetent E. coli are supplied as 10 x 100 L aliquots, with CopyControl™ Induction Solution and pUC19 Control DNA (100 pg/L).