missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TransformAid Bacterial Transformation Kit

Numéro de catalogue. ferk2711
Change view
Click to view available options
Quantity:
40 Transformations
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
FERK2711 40 Transformations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. FERK2711 Fournisseur Thermo Scientific™ Code fournisseur K2711
Il en reste null

Kit for quick and convenient preparation of competent E.coli cells for guaranted transformation efficiencies of more than 107 transformants per μg of plasmid DNA.

Employ a novel method for rapid preparation of chemically-competent E. coli cells from overnight bacterial cultures or bacterial colonies with the Thermo Scientific TransformAid™ Bacterial Transformation Kit. The key component of this system is the unique T-solution, which produces competent cells in a few easy steps. The quick and convenient procedure guarantees transformation efficiencies of more than 107 transformants per μg of plasmid DNA, ideal for routine cloning experiments. The option to use bacterial colonies for preparation of competent cells adds flexibility to any cloning experiment.

The TransformAid Bacterial Transformation Kit can be used with most E. coli strains commonly used for cloning: JM107, JM109, XL1-Blue, SURE, TOPP2, W3110, NM527, AD494, CJ236, GM2163, C600, BL21, M15, NM522, HB101, ER2267.

Highlights

  • Efficient – more than 107 transformants per μg of plasmid DNA
  • Practical – E. coli competent cells can be prepared from an overnight culture or from bacterial colonies
  • Fast – less than 1 hour from the overnight bacterial culture to cell plating. Starting from bacterial colonies, the procedure requires only 2.5 hours
  • Easy – all procedures are performed on ice on your bench. Brief centrifugations are carried out in regular microcentrifuges

Applications

  • Routine cloning experiments
  • Blunt-end cloning
  • TA cloning

Includes

  • C-Medium
  • T-Solution (A)
  • T-Solution (B)
  • Detailed Protocol

Note

Competent cells prepared with TransformAid Bacterial Transformation Kit are suitable for direct-use only. Freezing down and storage at -70°C is not recommended.

Spécifications

Cell Type E. coli
Format Kit
Species E. coli
Efficiency ≥1 x 107;
For Use With (Application) Cloning
Product Line TransformAid
Product Type TransformAid Bacterial Transformation Kit
Quantity 40 Transformations

For Research Use Only. Not for use in diagnostic procedures.