Research Products Corporation Disposable Plastic Transfer Pipettes, Sterile, Graduated 3ml Large Bulb, 7ml Capacity, Individually Wrapped, 500 per Package
Numéro de catalogue. 501367948
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling.
Numéro de catalogue.
501367948
Fournisseur
Research Products CorporationCode fournisseur
1475121S
Plastic transfer pipettes are molded into one-piece units from low density polyethylene. They eliminate cross-contamination encountered with rubber bulbs and reduce exposure when working with hazardous or infectious samples. Ideal for all laboratory procedures that require fast and safe transferring of liquids. Pipettes can also be used as freeze vials or as storage and transport containers by merely heat sealing the tips closed after filling.