missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-Farnesol, 97%

Catalog No. AAL1434814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1434814 25 g
AAL1434803 1 g
AAL1434806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1434814 Supplier Thermo Scientific Chemicals Supplier No. L1434814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-28-5 | C15H26O | 222.37 g/mol

trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-Farnesol is an intermediate used in the biological synthesis of cholesterol from mevalonic acid. It is used in the perfume industry. Further, it is used in the preparation of terpenes such as squalenes, cembranoids and forskolin. It is also used in the synthesis of Cecropia juvenile hormone.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 106-28-5
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
MDL Number MFCD00002918
InChI Key CRDAMVZIKSXKFV-YFVJMOTDSA-N
Synonym farnesol, trans,trans-farnesol, e,e-farnesol, trans-farnesol, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-ol, 2-trans,6-trans-farnesol, 2e,6e-farnesol, all-trans-farnesol, polyprenol, farnesyl alcohol
PubChem CID 445070
ChEBI CHEBI:16619
IUPAC Name (2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CO

Specifications

Density 0.886
Boiling Point 137°C (3 mmHg)
Flash Point 96°C (204°F)
Refractive Index 1.49
Quantity 25 g
Beilstein 1723039
Sensitivity Light sensitive
Merck Index 14,3937
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 222.37
Percent Purity 97%
Chemical Name or Material trans,trans-Farnesol
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 225-004-1
RTECSNumber JR4979000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only