missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-Dibenzylideneacetone, 98%

Catalog No. AC409001000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409001000 100 g Glass bottle
AC409000050 5 g Glass bottle
AC409000250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC409001000 Supplier Thermo Scientific Chemicals Supplier No. 409001000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35225-79-7 | C17H14O | 234.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35225-79-7
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.30
MDL Number MFCD00004790
InChI Key WMKGGPCROCCUDY-PHEQNACWSA-N
Synonym dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one
PubChem CID 640180
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
SMILES O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 107°C to 114°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula C6H5CH=CHC(O)CH=CHC6H5
Quantity 100 g
Beilstein 07, IV, 1747
Merck Index 15, 3010
Solubility Information Solubility in water: negliglible. Other solubilities: soluble in acetone and chloroform,slightly soluble in alcohol and ether
Formula Weight 234.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material trans, trans-Dibenzylideneacetone, 95%
Show More Show Less

RUO – Research Use Only