missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 71°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 302.48 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans,trans-4-(4-Fluorophenyl)-4′-propylbicyclohexyl |
Safety and Handling
| TSCA | No |