missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-2,4-Decadienal 90.0+%, TCI America™

Catalog No. D19345ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19345ML 5 mL
D193425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19345ML Supplier TCI America Supplier No. D19345ML

Chemical Identifiers

CAS 25152-84-5
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00007007
InChI Key JZQKTMZYLHNFPL-BLHCBFLLSA-N
Synonym 2e,4e-deca-2,4-dienal, 2,4-decadienal, e,e-2,4-decadienal, trans,trans-2,4-decadienal, 2,4-decadienal, 2e,4e, 2e,4e-decadienal, 2e,4e-2,4-decadienal, 2,4-decadienal, e,e, 2-trans-4-trans-decadienal, ccris 4029
PubChem CID 5283349
IUPAC Name (2E,4E)-deca-2,4-dienal
SMILES CCCCCC=CC=CC=O

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 mL
Formula Weight 152.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material trans,trans-2,4-Decadienal
RTECSNumber HD3000000
TSCA Yes