missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-1,5-Diphenyl-1,4-pentadien-3-one 99.0+%, TCI America™

Catalog No. p-7126494
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D090325G 25 g
D0903250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D090325G Supplier TCI America Supplier No. D090325G

Chemical Identifiers

CAS 35225-79-7
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.30
MDL Number MFCD00004790
InChI Key WMKGGPCROCCUDY-PHEQNACWSA-N
Synonym dibenzylideneacetone, 1,5-diphenylpenta-1,4-dien-3-one, trans,trans-dibenzylideneacetone, dibenzalacetone, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one, dibenzylidene acetone, distyryl ketone, 1,5-diphenyl-1,4-pentadien-3-one, trans,trans-dibenzalacetone, trans,trans-1,5-diphenyl-1,4-pentadien-3-one
PubChem CID 640180
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
SMILES O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 113°C
Quantity 25 g
Formula Weight 234.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,5-Diphenyl-1,4-pentadien-3-one