missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamic Acid Zone Refined (number of passes:40) 99.8+%, TCI America™

Catalog No. C0636ZONE
Change view
Click to view available options
Quantity:
ZONE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0636ZONE ZONE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0636ZONE Supplier TCI America Supplier No. C06361SAMPLE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 140-10-3
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00004369
InChI Key WBYWAXJHAXSJNI-VOTSOKGWSA-N
Synonym cinnamic acid, trans-cinnamic acid, e-cinnamic acid, 3-phenylacrylic acid, trans-3-phenylacrylic acid, zimtsaeure, 2e-3-phenylprop-2-enoic acid, 3-phenylprop-2-enoic acid, 3-phenylpropenoic acid
PubChem CID 444539
ChEBI CHEBI:35697
IUPAC Name (E)-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 135°C
Color White
Boiling Point 300°C
Quantity ZONE
Formula Weight 148.16
Percent Purity ≥99.8% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-Cinnamic Acid Zone Refined (number of passes:40)
EINECSNumber (3)-1719
RTECSNumber GD7850000
TSCA Yes