missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamaldehyde 98.0+%, TCI America™

Catalog No. C0352500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0352500ML 500 mL
C035225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0352500ML Supplier TCI America Supplier No. C0352500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14371-10-9
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007000
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
IUPAC Name (2E)-3-phenylprop-2-enal
SMILES O=C\C=C\C1=CC=CC=C1

Specifications

Melting Point -8°C
Color Yellow
Density 1.05 g/ml
Boiling Point 252°C
Quantity 500 mL
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-Cinnamaldehyde
EINECSNumber (3)-1148
RTECSNumber GD6476000
TSCA Yes