missing translation for 'onlineSavingsMsg'
Learn More

trans-Chalcone, 97%

Catalog No. AAA1473422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1473422 100 g
AAA1473436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1473422 Supplier Thermo Scientific Chemicals Supplier No. A1473422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-47-1 | C15H12O | 208.26 g/mol

Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trans chalcone is used to prevent lung and forestomach cancer. It induces programmed cell death due to reduced mitochondrial transmembrane potential in Arabidopsis thaliana roots.

Solubility
Soluble in chloroform, ether, benzene, and ethanol (slightly). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 614-47-1
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003082
InChI Key DQFBYFPFKXHELB-VAWYXSNFSA-N
Synonym chalcone, trans-chalcone, e-chalcone, benzalacetophenone, chalkone, benzylideneacetophenone, phenyl styryl ketone, cinnamophenone, 2-benzalacetophenone, 2-benzylideneacetophenone
PubChem CID 637760
ChEBI CHEBI:48965
IUPAC Name (E)-1,3-diphenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 55°C to 57°C
Boiling Point 208°C (25 mmHg)
Quantity 100 g
Beilstein 509985
Merck Index 14,2037
Solubility Information Soluble in chloroform,ether,benzene,and ethanol (slightly). Insoluble in water.
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material trans-Chalcone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-383-8
RTECSNumber UD5576750
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only