missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Styrylboronic acid, 97%

Catalog No. p-7035510
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5322703 1 g
AAH5322706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5322703 Supplier Thermo Scientific Chemicals Supplier No. H5322703

CAS: 6783-05-7 | C8H9BO2 | 147.97 g/mol

trans-beta-Styrylboronic acid as reagent is used for palladium (Pd)-catalyzed Suzuki-Miyaura coupling reactions, diastereoselective synthesis via Pd-catalyzed Heck-Suzuki cascade reaction and rhodium (Rh)-catalyzed intramolecular amination of aryl azides. It is also used as reagent in preparation of optically active unsaturated amino acids by diastereoselective Petasis borono-Mannich reaction and amino alcohol dienes via Petasis 3-component reaction using Ru-catalyzed ring-closing metathesis and isomerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-beta-Styrylboronic acid as reagent is used for palladium (Pd)-catalyzed Suzuki-Miyaura coupling reactions, diastereoselective synthesis via Pd-catalyzed Heck-Suzuki cascade reaction and rhodium (Rh)-catalyzed intramolecular amination of aryl azides. It is also used as reagent in preparation of optically active unsaturated amino acids by diastereoselective Petasis borono-Mannich reaction and amino alcohol dienes via Petasis 3-component reaction using Ru-catalyzed ring-closing metathesis and isomerization.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 6783-05-7
MDL Number MFCD00963621

Specifications

Melting Point 98°C to 100°C
Molecular Formula C11H10N4O4
Quantity 1 g
Synonym carbadox, unii-m2x04r2e2y, getroxel, mecadox, dsstox_cid_23913, dsstox_rid_80088, dsstox_gsid_43913, fortigro, karbadox czech, methyl n-e-1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene methyl iminocarbamate
InChI Key BPMVRAQIQQEBLN-UHFFFAOYSA-N
SMILES COC(=O)N=NC=C1C=[N+](C2=CC=CC=C2N1O)[O-]
IUPAC Name methyl N-[(E)-(1-hydroxy-4-oxidoquinoxalin-4-ium-2-ylidene)methyl]iminocarbamate
Molecular Weight (g/mol) 262.23
PubChem CID 5353472
Formula Weight 147.97
Percent Purity 97%
Chemical Name or Material trans-beta-Styrylboronic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only