missing translation for 'onlineSavingsMsg'
Learn More

trans-beta-Methylstyrene (stabilized with TBC) 97.0+%, TCI America™

Catalog No. M117510ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M117510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M117510ML Supplier TCI America Supplier No. M117510ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 873-66-5
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00009280
InChI Key QROGIFZRVHSFLM-QHHAFSJGSA-N
Synonym beta-methylstyrene, trans-beta-methylstyrene, 1-phenylpropene, isoallylbenzene, propenylbenzene, trans-1-phenyl-1-propene, 1-propenylbenzene, trans-propenylbenzene, 1-phenyl-1-propene, benzene, 1-propenyl
PubChem CID 252325
IUPAC Name [(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC=CC=C1

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 176°C
Quantity 10 mL
UN Number 3295
Formula Weight 118.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-beta-Methylstyrene (stabilized with TBC)
EINECSNumber (3)-0008
RTECSNumber DA8400500
TSCA No
Recommended Storage Refrigerator