missing translation for 'onlineSavingsMsg'
Learn More

trans-Aconitic acid, 98%

Catalog No. AAB2008714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2008714 25 g
AAB2008722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2008714 Supplier Thermo Scientific Chemicals Supplier No. B2008714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4023-65-8 | C6H6O6 | 174.108 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It plays an important role as an antimicrobial agent that blocks the transformation of Leishmania donovani amastigotes to prostigotes. trans-Aconitic acid is also used in synthesis of aconitate esters. It may be used for cross-linking polyvinyl alcohol film for preparing composite nanofiltration membranes.

Solubility
Soluble in water at 20°C 400g/L. Slightly soluble in ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 4023-65-8
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.108
MDL Number MFCD00002721
InChI Key GTZCVFVGUGFEME-HNQUOIGGSA-N
Synonym trans-aconitic acid, trans-aconitate, e-1-propene-1,2,3-tricarboxylic acid, aconitic acid, e-prop-1-ene-1,2,3-tricarboxylic acid, aconitic acid, trans, trans aconitic acid, e-aconitic acid, unii-7db37960cw, 1e-1-propene-1,2,3-tricarboxylic acid
PubChem CID 444212
ChEBI CHEBI:32806
IUPAC Name (E)-prop-1-ene-1,2,3-tricarboxylic acid
SMILES C(C(=CC(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point ∼190°C (decomposition)
Odor Odorless
Quantity 25 g
Merck Index 14,117
Solubility Information Soluble in water at 20°C 400g/L. Slightly soluble in ether.
Formula Weight 174.11
Percent Purity 98%
Chemical Name or Material trans-Aconitic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-688-6
RTECSNumber UD2380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only