missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methoxycinnamic acid, 98%

Catalog No. AAA1387030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1387030 250 g
AAA1387009 10 g
AAA1387018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1387030 Supplier Thermo Scientific Chemicals Supplier No. A1387030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 943-89-5 | C10H10O3 | 178.19 g/mol

Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.

Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 943-89-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00004398
InChI Key AFDXODALSZRGIH-QPJJXVBHSA-N
Synonym 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
PubChem CID 699414
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(\C=C\C(O)=O)C=C1

Specifications

Melting Point 171°C to 175°C
Quantity 250 g
Beilstein 510468
Solubility Information Soluble in ethanol,ethyl acetate and other organic solvents.
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material trans-4-Methoxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-405-4
RTECSNumber UD3391300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only