missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methoxy-beta-nitrostyrene, 98%

Catalog No. AAA1274522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274522 100 g
AAA1274506 5 g
AAA1274514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1274522 Supplier Thermo Scientific Chemicals Supplier No. A1274522

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5576-97-6 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5576-97-6
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00024826
InChI Key JKQUXSHVQGBODD-UHFFFAOYSA-N
Synonym 1-methoxy-4-2-nitrovinyl benzene, 4-methoxy-beta-nitrostyrene, p-2-nitrovinyl anisole, trans-4-methoxy-beta-nitrostyrene, e-1-methoxy-4-2-nitrovinyl benzene, 2'-nitro vinyl anisole, 1-methoxy-4-e-2-nitroethenyl benzene, 1-4-methoxyphenyl-2-nitroethene, 1-4-methoxyphenyl-2-nitroethylene, 4-2-nitrovinyl anisole
PubChem CID 697963
SMILES COC1=CC=C(C=C[N+]([O-])=O)C=C1

Specifications

Melting Point 86°C to 88°C
Quantity 100 g
Beilstein 2047935
IUPAC Name 1-methoxy-4-(2-nitroethenyl)benzene
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material trans-4-Methoxy-beta-nitrostyrene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only