missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Hydroxymethyl)cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. H12435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12435G 5 g
H12431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12435G Supplier TCI America Supplier No. H12435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66185-74-8
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD10566907,MFCD01734677,MFCD10566906
InChI Key VQMIUUBKKPIDBN-UHFFFAOYSA-N
PubChem CID 202819
IUPAC Name 4-(hydroxymethyl)cyclohexane-1-carboxylic acid
SMILES OCC1CCC(CC1)C(O)=O

Specifications

Melting Point 141°C
Quantity 5 g
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(Hydroxymethyl)cyclohexanecarboxylic Acid