missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline 99.0+%, TCI America™

Catalog No. H02965G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02965G 5 g
H029625G 25 g
H0296100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H02965G Supplier TCI America Supplier No. H02965G
Only null left

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00064320
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Specifications

Color White
Quantity 5 g
Formula Weight 131.13
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Hydroxy-L-proline
EINECSNumber (9)-1628
TSCA Yes