missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Bromocinnamaldehyde 98.0+%, TCI America™

Catalog No. p-7123479
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43371G 1 g
B43375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43371G Supplier TCI America Supplier No. B43371G

Chemical Identifiers

CAS 49678-04-8
Molecular Formula C9H7BrO
Molecular Weight (g/mol) 211.06
MDL Number MFCD03032509
InChI Key XYRAWLRFGKLUMW-UHFFFAOYSA-N
Synonym trans-3-(4-Bromophenyl)acrylaldehyde, trans-3-(4-Bromophenyl)-2-propenal, trans-3-(4-Bromophenyl)acrolein
PubChem CID 6285035
IUPAC Name 3-(4-bromophenyl)prop-2-enal
SMILES BrC1=CC=C(C=CC=O)C=C1

Specifications

Melting Point 80°C
Color Yellow
Quantity 1 g
Formula Weight 211.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Bromocinnamaldehyde