missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanol, 97%, Thermo Scientific™

Numéro de catalogue. p-4258746
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
100g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC346680050 5g Glass bottle
AC346680250 25g Glass bottle
AC346681000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC346680050 Fournisseur Thermo Scientific Chemicals Code fournisseur 346680050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 27489-62-9
Formule moléculaire C6H13NO
Poids moléculaire (g/mol) 115.17
Clé InChI IMLXLGZJLAOKJN-UHFFFAOYSA-N
Synonyme trans-4-aminocyclohexanol, 4-aminocyclohexanol, cis-4-aminocyclohexanol, cis-4-amino-cyclohexanol, cyclohexanol, 4-amino-, trans, 1r,4r-4-aminocyclohexan-1-ol, cyclohexanol, 4-amino, trans-4-hydroxycyclohexylamine, 1r,4r-4-aminocyclohexanol, aminocyclohexanol trans-4
PubChem CID 81293
Nom de l’IUPAC 4-aminocyclohexan-1-ol
SOURIRES C1CC(CCC1N)O

Spécifications

Nom ou substance chimique trans-4-Aminocyclohexanol
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min. (GC)
Emballage Glass bottle
Quantité 5g
Poids de formule 115.17
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-492-8

RUO – Research Use Only