missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(3,4-Difluorophenyl)-trans-4'-ethylbicyclohexane 97.0+%, TCI America™

Numéro de catalogue. D38561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D38561G 1 g
D38565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D38561G Fournisseur TCI America Code fournisseur D38561G
Il en reste null

Identifiants chimiques

CAS 118164-50-4
Molecular Formula C20H28F2
Molecular Weight (g/mol) 306.441
MDL Number MFCD11053389
InChI Key WDECTZREPSJUQW-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-ethyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-ethyl-bicyclohexyl, 4-4-4-ethylcyclohexyl cyclohexyl-1,2-difluorobenzene, trans,trans-4-3,4-difluorophenyl-4-ethylbicyclohexyl, trans-4-3,4-difluorophenyl-trans-4'-ethylbicyclohexane, 4-trans-4-trans-4-ethylcyclohexyl cyclohexyl-1,2-difluorobenzene, 4-3,4-difluorophenyl-4'-ethyl-1,1'-bi cyclohexane, benzene, 4-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro, benzene, 4-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro, 4-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro-benzene
PubChem CID 550421
IUPAC Name 4-[4-(4-ethylcyclohexyl)cyclohexyl]-1,2-difluorobenzene
SMILES CCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Spécifications

Melting Point 53°C
Color White
Quantity 1 g
Formula Weight 306.44
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(3,4-Difluorophenyl)-trans-4′-ethylbicyclohexane
EINECSNumber (4)-1823
TSCA No