missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexène 99,0+%, TCI America™

Numéro de catalogue. H03991ML
Modifier l'affichage
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H03991ML 1 mL
H03995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H03991ML Fournisseur TCI America Code fournisseur H03991ML

Identifiants chimiques

CAS 13269-52-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009386
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
Synonym trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
PubChem CID 638066
IUPAC Name (E)-hex-3-ène
SMILES CCC=CCC

Spécifications

Color Incolore
Boiling Point 67°C
Quantity 1 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥99.0% (GC)
Physical Form Liquide
Chemical Name or Material trans-3-Hexene
EINECSNumber (2)-0031
TSCA No